missing translation for 'onlineSavingsMsg'
Learn More

Suramin hexasodium salt, 98+%

Catalog No. aaj61707md
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61707MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61707MD Supplier Thermo Scientific Chemicals Supplier No. J61707MD

Suramin hexasodium salt (suramin sodium, CAS # 129-46-4) is the sodium salt of suramin. | CAS: 129-46-4 | C51H34N6Na6O23S6 | 1429.15 g/mol

Suramin Sodium has been observed as a potent inhibitor of melanoma HPA (heparanase) and tumor cell metastasis. Inhibition of NAD+-dependent deacetylase SIRT1 with an IC50 of 2.6 ?M along with SIRT5 has also been observed. Suramin Sodium has been recorded to antagonize EDG-3 (S1P3) selectively. Suramin Sodium is an inhibitor of A cyclase, FGF-1, FGF-2, IL-1, IL-4, PDGF, PC-PLD, PKC, SH-PTP, TERT, TGF beta 1, Topo I, Topo II and VEGF. It is used as a G protein uncoupler and inhibitor of PDGF, EGF, TGF-β, SIRT1 and SIRT5. It also blocks calmodulin binding to recognition sites and G protein coupling to G protein-coupled receptors. Increases open probability of ryanodine receptor (RyR) channels. Anticancer and antiviral agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Suramin is a polysulphonated napthylurea which exhibits anti-parasitic and anticancer properties

• Suramin prevents endothelial cell migration and proliferation through inhibition of IGF-I, EGF, PDGF, and TGF-beta

• Suramin can also uncouple G-proteins from receptors and turn off cell signaling cascades

Application

• Suramin inhibits angiogenesis through blockade of bFGF and VEGF and can be useful for the laboratory study of blood vessel growth and renewal following injury

• Suramin inhibits DNA topoisomerases, and can be used as an antineoplastic agent in the laboratory study of cancer or for studies of microbial physiology

Specifications

CAS 129-46-4
Color White
Molecular Formula C51H34N6Na6O23S6
Quantity 250 mg
MDL Number MFCD00210217
Beilstein 3694087
Merck Index 14,9006
Synonym suramin sodium, suramin hexasodium, suramin hexasodium salt, suramin sodium salt, antrypol, moranyl, naganinum, naphuride sodium, fourneau 309, suramine sodium salt
Solubility Information Soluble in water and saline; Sparingly soluble in 95% bethanol
InChI Key VAPNKLKDKUDFHK-UHFFFAOYSA-H
SMILES [Na+].[Na+].[Na+].[Na+].[Na+].[Na+].CC1=CC=C(C=C1NC(=O)C1=CC(NC(=O)NC2=CC(=CC=C2)C(=O)NC2=CC(=CC=C2C)C(=O)NC2=C3C(C=C(C=C3S([O-])(=O)=O)S([O-])(=O)=O)=C(C=C2)S([O-])(=O)=O)=CC=C1)C(=O)NC1=C2C(C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O)=C(C=C1)S([O-])(=O)=O
IUPAC Name hexasodium 8-{4-methyl-3-[3-({[3-({2-methyl-5-[(4,6,8-trisulfonatonaphthalen-1-yl)carbamoyl]phenyl}carbamoyl)phenyl]carbamoyl}amino)benzamido]benzamido}naphthalene-1,3,5-trisulfonate
Molecular Weight (g/mol) 1429.15
PubChem CID 8514
Formula Weight 1429.16
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Suramin hexasodium salt
Show More Show Less
EINECSNumber 204-949-3
RTECSNumber QM7000000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only