missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperSignal™ West Femto and SuperSignal™ West Pico PLUS Sample

Catalog No. pia43841
Change view
Click to view available options
Quantity:
200 mL plus 20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA43841 200 mL plus 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIA43841 Supplier Thermo Scientific™ Supplier No. A43841

Provides chemiluminescent detection substrates for both routine and high sensitivity western blots

SuperSignal™ West Pico PLUS:

  • SuperSignal™ West Pico PLUS Chemiluminescent Substrate enables picogram to high femtogram-level protein detection by western blot analysis
  • It is best for routine, everyday western blotting applications, providing excellent signal intensity, sensitivity and duration across a wide range of targets
  • Sensitivity: pictogram to femtogram sensitivity
  • Stability: 8-hr working solution stability; 1-year kit stability at room temperature
  • Compatibility: nitrocellulose and PVDF membranes
  • Signal duration: 6 to 24 hours
  • Recommended primary antibody concentration: 1:1,000–1:5,000 dilution (0.2–1.0 μg/mL)
  • Recommended secondary antibody concentration: 1:20,000–100,000 dilution (10–50 ng/mL)

SuperSignal™ West Femto:

  • SuperSignal™ West Femto Chemiluminescent Substrate is an ultra-sensitive enhanced chemiluminescent (ECL) substrate for low femtogram protein level detection by western blot analysis with horseradish peroxidase (HRP) enzyme
  • It is optimized to detect low abundant and/or difficult to detect targets
  • Sensitivity: low-femtogram (mid-zeptomole)
  • Stability: 8-hr working solution stability, 1-year kit stability at 4°C
  • Compatibility: nitrocellulose and PVDF membranes
  • Signal duration: up to 8 hours
  • Recommended primary antibody concentration: 1:5,000–1:100,000 dilution (10–200 ng/mL)
  • Recommended secondary antibody concentration: 1:100,000–1:500,000 dilution (2–10 ng/mL)

Specifications

Product Type HRP Substrate
For Use With (Application) Western Blot
Quantity 200 mL plus 20 mL
Content And Storage Room Temperature and 4°C