missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperSignal™ West Femto Maximum Sensitivity Substrate

Ultra-sensitive enhanced chemiluminescent (ECL) HRP substrate for low-femtogram-level detection by Western blot analysis.

Supplier:  Thermo Scientific™ 34096X4

 View more versions of this product

Catalog No. FER34096X4


Only null left

Description

Description

Thermo Scientific™ SuperSignal West Femto Substrate is an ultra-sensitive enhanced chemiluminescent substrate (ECL substrate) for detection of peroxidase activity from HRP-conjugated antibodies and probes in Western blotting.

SuperSignal West Femto Substrate Maximum Sensitivity Substrate is an ultra-sensitive chemiluminescent substrate system for Western blotting with horseradish peroxidase (HRP) enzyme. The substrate kit enables detection of low femtogram (that's mid-zeptomole!) amounts of protein deposited on nitrocellulose or PVDF membrane and probed with appropriate primary and secondary antibodies. When combined with optimized antibody concentrations and blocking buffers, SuperSignal West Femto Substrate enables detection of target proteins in amounts that are small to be seen with typical ECL substrates.

Highlights:

  • ECL substrate – an enhanced chemiluminescent substrate for detection of horseradish peroxidase (HRP) activity from antibodies and other Western blot probes
  • Sensitive – detect low-femtogram (mid-zeptomole) amounts of protein in bands on nitrocellulose or PVDF membranes
  • Quantitative – produces quantitative signal that is measurable over two orders of magnitude
  • Saves on antibody – optimized for very dilute antibody concentrations; dilute primary antibodies 5000- to 100,000-fold and secondary antibodies 100,000 to 500,000-fold (from 1 mg/mL stocks)
  • Intense signal – easy to capture an image by exposure to autoradiography film or via a CCD camera or similar imaging system
  • Excellent signal duration – 8 hours of usable light-output when conditions are optimized
  • Stable reagent – kit components are stable for 1 year at 4°C or 6 months at room temperature
Specifications

Chemical Identifiers

Substrate
Liquid
Horseradish Peroxidase
Chemiluminescence
SuperSignal
6 to 8 hr.

Specifications

Substrate
Liquid
4 x 200 mL
Horseradish Peroxidase
Low to mid-femtogram
Luminol/Enhancer, 4 x 100 mL, Stable Peroxide Buffer, 4 x 100 mL
Chemiluminescence
SuperSignal
HRP
2°: 1:100-500 K (2-10 ng/mL), 1°: 1:5 K (10- 200 ng/mL)
6 to 8 hr.
Sufficient For: 100 mini-gel blots; 8000 cm2 of membrane
• Luminol/Enhancer, 4 x 100 mL
• Stable Peroxide Buffer, 4 x 100 mL

Store reagents at 4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.