missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperSignal™ West Atto Ultimate Sensitivity Substrate

SuperSignal™ West Atto Ultimate Sensitivity Substrate is an enhanced chemiluminescent (ECL) substrate for high attogram protein detection and provides the high level sensitivity and better signal-to-noise ratios compared to other commercially available horseradish peroxidase (HRP) substrates.

Supplier:  Thermo Scientific™ A38556

 View more versions of this product

Catalog No. PIA38556


Only null left
Explore available promotions

Description

Description

  • Sensitivity: low-femtogram to high-attogram
  • Stability: eight hours for working solution, one year for kit at 4°C
  • Compatibility: nitrocellulose and PVDF membranes
  • Signal duration: up to four hours
  • Recommended primary antibody concentration: 1:1000 to 1:5000 dilution (0.2 to 1.0 μg/mL)
  • Recommended secondary antibody concentration: 1:100000 to 1:250000 dilution (4 to 10 ng/mL)
Specifications

Specifications

Substrate
Liquid
Nitrocellulose, PVDF
200 mL
Low-femtogram to high-attogram
Luminol/Enhancer, 100 mL, Stable Peroxide Buffer, 100 mL
Wet Ice
Chemiluminescence
Western Blot
SuperSignal
Horseradish Peroxidase
25 Mini-Gel Blots, 2000 sq. cm of Membrane
SuperSignal West Atto Substrate / SuperSignal West Atto Stable Peroxide

• Substrate Solution, 100 mL
• Stable Peroxide Solution, 100 mL

Sufficient for 25 mini-gel blots; 2000 cm2 of membrane.

Store reagents at 2–8°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.