missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperSignal™ West Atto Ultimate Sensitivity Substrate

Catalog No. pia38554
Change view
Click to view available options
Quantity:
20 mL
100 mL
200 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA38554 20 mL
PIA38555 100 mL
PIA38556 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PIA38554 Supplier Thermo Scientific™ Supplier No. A38554
Only null left

SuperSignal™ West Atto Ultimate Sensitivity Substrate is an enhanced chemiluminescent (ECL) substrate for high attogram protein detection and provides the high level sensitivity and better signal-to-noise ratios compared to other commercially available horseradish peroxidase (HRP) substrates.

  • Sensitivity: low-femtogram to high-attogram
  • Stability: eight hours for working solution, one year for kit at 4°C
  • Compatibility: nitrocellulose and PVDF membranes
  • Signal duration: up to four hours
  • Recommended primary antibody concentration: 1:1000 to 1:5000 dilution (0.2 to 1.0 μg/mL)
  • Recommended secondary antibody concentration: 1:100000 to 1:250000 dilution (4 to 10 ng/mL)

Specifications

Product Type Substrate
Detection Method Chemiluminescence
Form Liquid
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Product Line SuperSignal
Quantity 20 mL
Substrate Horseradish Peroxidase
Sensitivity Low-femtogram to high-attogram
Sufficient For 2 Mini-Gel Blots, 200 sq. cm of Membrane
Kit Contents Luminol/Enhancer, 10 mL, Stable Peroxide Buffer, 10 mL
Content And Storage SuperSignal West Atto Substrate / SuperSignal West Atto Stable Peroxide

• Substrate Solution, 10 mL
• Stable Peroxide Solution, 10 mL

Sufficient for two mini-gel blots; 200 cm2 of membrane.

Store reagents at 2–8°C.
Shipping Condition Wet Ice
Show More Show Less
How do I handle saturated or burned out bands using SuperSignal West Atto Ultimate Sensitivity Substrate?

Here are some suggestions:
- Shorten the time of exposure
- Wash blot and try using SuperSignal West Pico PLUS Chemiluminescent Substrate
- Strip the membrane using Restore Western Blot Stripping Buffer, re-probe with lesser amount of primary and secondary antibodies, and try first with SuperSignal West Pico PLUS Chemiluminescent Substrate
- Re-run the gel using lesser sample

How can I reduce background with SuperSignal West Atto Ultimate Sensitivity Substrate?

Here are potential causes for high background and corresponding solutions:
- Too much HRP in the system: Further dilute the HRP-conjugate, see guidelines in Table 1 in the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018858_SuperSignalWestAtto_UG.pdf).
- Inadequate blocking: Optimize blocking conditions.
- Inappropriate blocking reagent: Try a different blocking reagent. Use clear milk or StartingBlock blocking buffer instead of dry milk.
- Inadequate washing: Increase length, number or volume of washes.
- Film has been overexposed: Decrease exposure time or use Pierce Background Eliminator (Cat No. 21065).
- Concentration of antigen or antibody is too high: Decrease amount of antigen or antibody.
- Poor antibody specificity: Use SuperSignal Western Blot Enhancer (Cat. No. 46640).
We also recommend minimizing handling of the membrane, using clean gloves while handling, clean plasticware, glassware, forceps, or other contact surfaces. We also recommend trimming of the membrane to remove excess area.

Is SuperSignal West Atto Ultimate Sensitivity Substrate compatible with Restore Western Blot Stripping Buffer?

SuperSignal West Atto Ultimate Sensitivity Substrate is compatible with Restore Western Blot Stripping Buffer.

When should I use SuperSignal West Atto Ultimate Sensitivity Substrate?

We recommend using SuperSignal West Atto Ultimate Sensitivity Substrate for detection of proteins in the following situations:
- For proteins with low concentration, not measurable by BCA or Bradford assays
- For low-abundance phosphorylated or glycosylated proteins
- For low-abundance membrane or nuclear proteins
- For low-abundance proteins being detected with a low-affinity antibody or when the antibody is precious
- When sample source is limited or when multiple conditions or assays are being tested on a single sample

Is SuperSignal West Atto Ultimate Sensitivity Substrate affected by freeze/thaw cycles?

SuperSignal West Atto Ultimate Sensitivity Substrate can tolerate up to 3 freeze/thaw cycles without significant loss in performance.

What is the detection range for SuperSignal West Atto Ultimate Sensitivity Substrate?

SuperSignal West Atto Ultimate Sensitivity Substrate is an ultra-sensitive enhanced chemiluminescent (ECL) substrate that can detect low-femtogram to high-attogram quantities of protein.


For Research Use Only. Not for use in diagnostic procedures.