Learn More
Invitrogen™ SuperScript™ IV RT-LAMP Master Mix
Description
Invitrogen SuperScript IV RT-LAMP Master Mix is a ready-to-use solution optimized for fast, sensitive, and simple RNA target detection in loop-mediated isothermal amplification (LAMP) reactions. With this convenient master mix formulation, you can perform isothermal amplification from both RNA or DNA targets in a single tube format.
Benefits of SuperScript IV RT-LAMP Master Mix
- RNA and DNA target detection using Bst DNA polymerase for isothermal amplification in as little as five minutes
- Robust and specific cDNA synthesis using SuperScript IV Reverse Transcriptase
- Enhanced specificity and sensitivity with buffer formulation and RNaseOUT recombinant ribonuclease inhibitor
- Compatible with real-time and endpoint detection—includes SYTO 9 dye for real-time detection
- Flexible design provides several options for evaluating results, including real-time and endpoint detection methods
SuperScript IV RT-LAMP Master Mix includes an evolved Bst DNA polymerase mutant with a fast reaction speed and increased sensitivity and tolerance to inhibitors, as well as SuperScript IV Reverse Transcriptase with increased processivity, thermostability, and inhibitor resistance. To prevent RNA degradation by carry-over RNases, the master mix includes the potent, noncompetitive RNaseOUT Recombinant Ribonuclease Inhibitor. Our proprietary reaction buffer composition is optimized to increase specificity by delaying no-template control (NTC) amplification time.
Specifications
Specifications
| Product Type | Master Mix |
| For Use With (Application) | LAMP, RT-LAMP |
| Polymerase | Bst DNA Polymerase |
| Quantity | 1000 reactions, 10 x 1.25 mL |
| Content And Storage | • 2X SuperScript IV RT-LAMP Master Mix (10 x 1.25 mL) |
| No. of Reactions | 1000 Reactions |
| Optimal Reaction Temperature | 65°C |
| Reverse Transcriptase | SuperScript IV |
| Shipping Condition | Dry Ice |
| Detection Method | Real-time and End-point Detection |
| Show More |
Frequently Asked Questions (FAQs)
Yes, LAMP can be combined with colorimetric detection using calcein. We recommend adding calcein and MnCl₂ to the reaction mixture at final concentrations of 80 µM and 0.25 mM, respectively. Prior to amplification, calcein binds to Mn²⁺ ions, giving the mixture an orange color. During amplification, Mn²⁺ ions form a complex with pyrophosphate, releasing calcein and causing the reaction mixture to change to a green or yellowish-green color, indicating successful amplification.