missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ IV RT-LAMP Master Mix

Catalog No. A51801
Encompass
Change view
Click to view available options
No. of Reactions:
100 Reactions
1000 Reactions
400 Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
A51801 100 Reactions
A51802 400 Reactions
A51803 1000 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A51801 Supplier Invitrogen™ Supplier No. A51801

Invitrogen SuperScript IV RT-LAMP Master Mix is a ready-to-use solution optimized for fast, sensitive, and simple RNA target detection in loop-mediated isothermal amplification (LAMP) reactions.

Invitrogen SuperScript IV RT-LAMP Master Mix is a ready-to-use solution optimized for fast, sensitive, and simple RNA target detection in loop-mediated isothermal amplification (LAMP) reactions. With this convenient master mix formulation, you can perform isothermal amplification from both RNA or DNA targets in a single tube format. 

Benefits of SuperScript IV RT-LAMP Master Mix
• RNA and DNA target detection using Bst DNA polymerase for isothermal amplification in as little as five minutes
• Robust and specific cDNA synthesis using SuperScript IV Reverse Transcriptase
• Enhanced specificity and sensitivity with buffer formulation and RNaseOUT recombinant ribonuclease inhibitor
• Compatible with real-time and endpoint detection—includes SYTO 9 dye for real-time detection
• Flexible design provides several options for evaluating results, including real-time and endpoint detection methods

SuperScript IV RT-LAMP Master Mix includes an evolved Bst DNA polymerase mutant with a fast reaction speed and increased sensitivity and tolerance to inhibitors, as well as SuperScript IV Reverse Transcriptase with increased processivity, thermostability, and inhibitor resistance. To prevent RNA degradation by carry-over RNases, the master mix includes the potent, noncompetitive RNaseOUT Recombinant Ribonuclease Inhibitor. Our proprietary reaction buffer composition is optimized to increase specificity by delaying no-template control (NTC) amplification time.

Specifications

Product Type Master Mix
For Use With (Application) LAMP, RT-LAMP
Polymerase Bst DNA Polymerase
Quantity 100 reactions, 1 x 1.25 mL
Content And Storage

• 2X SuperScript IV RT-LAMP Master Mix (1.25 mL)
• SYTO Green Fluorescent Nucleic Acid Stain (30 μL)

Store at –20°C and protect from light.

No. of Reactions 100 Reactions
Optimal Reaction Temperature 65°C
Reverse Transcriptase SuperScript IV
Shipping Condition Dry Ice
Detection Method Real-time and end-point detection
Includes Label or Dye SYTO 9 Green Fluorescent Nucleic Acid Stain
Concentration 2X
Reaction Time 5 to 15 minutes
Show More Show Less
Is it possible to combine an (RT-)LAMP reaction with colorimetric detection methods using SuperScript IV RT-LAMP Master Mix?

Yes, LAMP can be combined with colorimetric detection using calcein. We recommend adding calcein and MnCl₂ to the reaction mixture at final concentrations of 80 µM and 0.25 mM, respectively. Prior to amplification, calcein binds to Mn²⁺ ions, giving the mixture an orange color. During amplification, Mn²⁺ ions form a complex with pyrophosphate, releasing calcein and causing the reaction mixture to change to a green or yellowish-green color, indicating successful amplification.