Learn More
Superior Glove™ Superior Touch™ Cleanroom Processed Cut-Resistant Gloves
Dyneema™ fiber gloves with polyurethane coated palms are clean processed and ideal for sharp steel, glass, food and circuit board applications.
Supplier: Superior Glove™ CS13SXPUQ7
Description
Dyneema™ fiber gloves with polyurethane coated palms are clean processed and ideal for sharp steel, glass, food and circuit board applications.
- Dyneema™ is lint-free and provides cut and abrasion resistance
- Polyurethane coated palm provides secure grip in wet and oily conditions
- Form fitting, breathable and flexible
- Ergonomically shaped
Specifications
| Polyurethane | |
| ANSI A2 | |
| Cut-resistant Gloves | |
| Sharp steel, glass, circuit boards, food applications | |
| CS13SXPUQ-7 | |
| Superior Touch™ |
| White | |
| Dyneema | |
| 7 | |
| Knit | |
| ANSI/ISEA 105-16 Level 3 |