missing translation for 'onlineSavingsMsg'
Learn More

Superior Glove™ Superior Touch™ Cleanroom Processed Cut-Resistant Gloves

Catalog No. p-4408806
Change view
Click to view available options
Size:
6
7
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17489646 6
17489647 7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17489646 Supplier Superior Glove™ Supplier No. CS13SXPUQ6
Only null left

Dyneema™ fiber gloves with polyurethane coated palms are clean processed and ideal for sharp steel, glass, food and circuit board applications.

  • Dyneema™ is lint-free and provides cut and abrasion resistance
  • Polyurethane coated palm provides secure grip in wet and oily conditions
  • Form fitting, breathable and flexible
  • Ergonomically shaped

Specifications

Coating Material Polyurethane
Color White
Cut Resistance Rating ANSI A2
Material Dyneema
Product Type Cut-resistant Gloves
Size 6
For Use With (Application) Sharp steel, glass, circuit boards, food applications
Glove Cuff Style Knit
Model CS13SXPUQ-6
Puncture Rating ANSI/ISEA 105-16 Level 3
Series Superior Touch™
Show More Show Less