Learn More
Superior Glove™ Chemstop™ Fully Nitrile Coated Cotton Gloves
Nitrile rubber coating is resistant to oils and grease for enhanced grip capabilities.
Supplier: Superior Glove™ GNTFC9
Description
- Nitrile rubber coating covers the entire glove surface
- Gloves are resistant to grease and oil for enhanced grip capabilities
- Knit wrist prevents dirt and debris from entering the glove
Specifications
| ANSI 3 | |
| Blue, White | |
| Dip-coated Gloves | |
| Brick Laying, Castings, Metal Stamping, Sheet Metal, and Plywood | |
| ANSI/ISEA 105-16 Level 2 |
| Nitrile | |
| Specific | |
| 9 | |
| Knit | |
| GNTFC |