missing translation for 'onlineSavingsMsg'
Learn More

Superior Glove™ Endura™ Goatskin Leather Drivers Gloves with Keystone Thumb

Goatskin is often referred to as nature's strongest leather due to its high tensile-strength and abrasion-resistant properties.

$112.39 - $141.78

Specifications

Color White
Hand Specific
Material Goatskin Leather
Product Type Driver Gloves
For Use With (Application) Janitorial, Material Handling, General Maintenance, Construction, Welding, and Agriculture
View More Specs

Products 6
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
29-635-7084
View Documents
Superior Glove™
378GKTAXL
X-Large
Dozen of 12 Pair for $112.39
Case of 10 Dozen for $1,123.90
Only null left
 
29-636-2075
View Documents
Superior Glove™
378GKTA2XL
2X-Large
Dozen of 12 Pair for $141.78
Case of 10 Dozen for $431.60
Only null left
 
29-636-4131
View Documents
Superior Glove™
378GKTAXS
X-Small
Dozen of 12 Pair for $112.39
Case of 10 Dozen for $1,123.90
Only null left
 
29-635-7081
View Documents
Superior Glove™
378GKTAS
Small
Dozen of 12 Pair for $112.39
Case of 10 Dozen for $1,123.90
Only null left
 
29-635-7082
View Documents
Superior Glove™
378GKTAM
Medium
Dozen of 12 Pair for $112.39
Case of 10 Dozen for $345.30
Only null left
 
29-635-7083
View Documents
Superior Glove™
378GKTAL
Large
Dozen of 12 Pair for $112.39
Case of 10 Dozen for $345.30
Only null left
 
Description

Description

  • Goatskin leather
  • High tensile-strength
  • Abrasion resistant
  • Keystone thumb
Specifications

Specifications

White
Goatskin Leather
Janitorial, Material Handling, General Maintenance, Construction, Welding, and Agriculture
Yellow
Keystone
Specific
Driver Gloves
Open
378GKTA
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'