missing translation for 'onlineSavingsMsg'
Learn More

Superior Glove™ Endura™ Goatskin Leather Drivers Gloves with Keystone Thumb

Catalog No. 296357084
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296357084 X-Large
296362075 2X-Large
296364131 X-Small
296357081 Small
296357082 Medium
296357083 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 296357084 Supplier Superior Glove™ Supplier No. 378GKTAXL

Goatskin is often referred to as nature's strongest leather due to its high tensile-strength and abrasion-resistant properties.

  • Goatskin leather
  • High tensile-strength
  • Abrasion resistant
  • Keystone thumb

Specifications

Color White
Hand Specific
Material Goatskin Leather
Product Type Driver Gloves
Size X-Large
For Use With (Application) Janitorial, Material Handling, General Maintenance, Construction, Welding, and Agriculture
Glove Cuff Style Open
Hem Color Yellow
Series 378GKTA
Thumb Style Keystone