Learn More
Superior Glove™ Dexterity™ Cut-Resistant String Knit Gloves, Nitrile Coating
Aramid fiber composite string knit gloves provides ANSI Level A3 cut-resistance. Nitrile coating makes this glove ideal for a variety of dry grip applications.
Supplier:  Superior Glove™ S13KFNT10
 View more versions of this product
Catalog No. 17-400-195
Only null left
Explore available promotions
Description
Description
  • Combination of Kevlar™ and composite filament fiber provides 360° cut protection
  • Nitrile palm coating offers secure dry grip and durability
  • Ergonomically designed for unsurpassed comfort and dexterity
  • Cut, abrasion and puncture resistant
Specifications
Specifications
ANSI 3
Cream, Black
Dexterity
10
Knit
ANSI/ISEA 105-16 Level 3
Nitrile Foam
ANSI A3
Cut-resistant Gloves
Automotive, light metal stamping, small parts handling, maintenance jobs, HVAC, glass
S13KFNT/10
Dexterity™
Product Suggestions
Product Suggestions