Learn More
Superior Glove™ Dexterity™ Nitrile Dipped Work Gloves
Seamless knit cotton shell is comfortable and flexible, with a special additive in the nitrile coating to make gloves 100% biodegradable.
Supplier: Superior Glove™ S15NT7
Description
- Nitrile dipped palm coating offers abrasion and puncture resistance
- An additive in the coating makes gloves 100% biodegradable
- Seamles knit cotton shell is comfortable and flexible
- CFIA compliant
Specifications
| ANSI 4 | |
| Nitrile | |
| Specific | |
| Dip-coated Gloves | |
| Automotive Assembly, Maintenance, and Metal Stamping | |
| ANSI/ISEA 105-16 Level 2 |
| Seamless Knit | |
| Blue, White | |
| Cotton | |
| 7 | |
| Knit | |
| S15NT |