Learn More
Superior Glove™ Dexterity™ 13-Gauge Nylon Gloves with Foam PVC Palms
Fine-gauge nylon construction is durable and abrasion resistant, while also increasing touch sensitivity. Foam PVC coating on the palm enhances grip capabilities in wet or oily conditions.
Supplier: Superior Glove™ S13BPVC7
Description
- PVC coating provides wear and abrasion resistance similar to that of nitrile, without the risk of glue adhering to its surface
- Fine-gauge nylon is durable, while also increasing touch sensitivity
- Enhanced grip in wet or oily conditions
- Silicone-free for use in the automotive industry
Specifications
| ANSI 3 | |
| PVC | |
| Specific | |
| Seamless Knit Gloves | |
| Small Parts Handling, Maintenance, Construction, and Automotive Assembly | |
| S13BPVC |
| Seamless Knit | |
| Blue | |
| Nylon | |
| 7 | |
| ANSI/ISEA 105-16 Level 2 | |
| 13 |