missing translation for 'onlineSavingsMsg'
Learn More

Superior Glove™ Dexterity™ 13-Gauge Nylon Gloves with Foam PVC Palms

Catalog No. p-4939937
Change view
Click to view available options
Size:
10
11
6
7
8
9
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296356932 11
296356931 10
296356930 9
296356929 8
296356927 6
296356928 7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 296356932 Supplier Superior Glove™ Supplier No. S13BPVC11
Only null left

Fine-gauge nylon construction is durable and abrasion resistant, while also increasing touch sensitivity. Foam PVC coating on the palm enhances grip capabilities in wet or oily conditions.

  • PVC coating provides wear and abrasion resistance similar to that of nitrile, without the risk of glue adhering to its surface
  • Fine-gauge nylon is durable, while also increasing touch sensitivity
  • Enhanced grip in wet or oily conditions
  • Silicone-free for use in the automotive industry

Specifications

ANSI Abrasion Rating ANSI 3
Construction Seamless Knit
Coating Material PVC
Color Blue
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size 11
For Use With (Application) Small Parts Handling, Maintenance, Construction, and Automotive Assembly
Puncture Rating ANSI/ISEA 105-16 Level 2
Series S13BPVC
Yarn Gauge 13
Show More Show Less