missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Superfrost™ Cygen™ Microscope Slide
Description
- For use with cytogenetic-specific applications
- Certified draw-line free glass surface
Specifications
Specifications
| No. per Pack | 144 |
| Material | Soda Lime Float Glass |
| Color | White |
| Description | Cytogenetic-specific slide |