missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Simport™ Scientific SuperFlex™ 'Break-Away' 96-well PCR Plates
Break-Away' PCR plates allow for flexible, efficient, and cost-effective PCR. Wells can be separated without scissors into 8 strips or higher, saving time and money.
Specifications
| Certifications/Compliance | RNase, DNase, Pyrogen and D-free |
|---|---|
| Certified | Yes |
| Format | Standard |
| Lid | Without Lid |
| Autoclavable | Autoclavable |
| Catalog Number | Mfr. No. | Color | Sterility | Fit for Purpose | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Sterility | Fit for Purpose | Price | Quantity & Availability | |||
|
23045580
|
Simport™ Scientific
T32096N |
Clear | Non-sterile | PCR Ready |
N/A
|
|
|||
|
23045581
|
Simport™ Scientific
T32096N1S |
Clear | Sterile | Certified Quality |
N/A
|
|
|||
|
23045582
|
Simport™ Scientific
T32096W |
White | Non-sterile | Certified Quality |
N/A
|
|
|||
Description
- Eliminates the use of scissors that cause perforated wells, damage sealing rings, and risk contamination issues
- Prevents need to throw away half-empty plates
- Compatible with PCR instruments that fit non-skirted, low profile plates
- Molded from virgin polypropylene
- Wells are individually flexible for perfect fit, even with a difference in expansion coefficients
- White model improves signal reflection, reduces variation, and increases sensitivity of qPCR assays
- Sealable with T321-96N PCR cap strips
Specifications
| RNase, DNase, Pyrogen and D-free | |
| Standard | |
| Autoclavable | |
| LoBind Skirted | |
| 0.2 mL | |
| PCR and qPCR |
| Yes | |
| Without Lid | |
| Single-use | |
| Polypropylene | |
| Conical Bottom (V-well) |