missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Simport™ Scientific SuperFlex™ 'Break-Away' 96-well PCR Plates
Break-Away' PCR plates allow for flexible, efficient, and cost-effective PCR. Wells can be separated without scissors into 8 strips or higher, saving time and money.
Supplier: Simport™ Scientific T32096W
Description
- Eliminates the use of scissors that cause perforated wells, damage sealing rings, and risk contamination issues
- Prevents need to throw away half-empty plates
- Compatible with PCR instruments that fit non-skirted, low profile plates
- Molded from virgin polypropylene
- Wells are individually flexible for perfect fit, even with a difference in expansion coefficients
- White model improves signal reflection, reduces variation, and increases sensitivity of qPCR assays
- Sealable with T321-96N PCR cap strips
Chemical Identifiers
| RNase, DNase, Pyrogen and D-free | |
| Standard | |
| Autoclavable | |
| LoBind Skirted | |
| 0.2 mL | |
| PCR and qPCR |
| Yes | |
| Without Lid | |
| Single-use | |
| Polypropylene | |
| Conical Bottom (V-well) |
Specifications
| RNase, DNase, Pyrogen and D-free | |
| Standard | |
| Autoclavable | |
| Single-use | |
| Polypropylene | |
| Non-sterile | |
| PCR and qPCR |
| Yes | |
| Without Lid | |
| White | |
| LoBind Skirted | |
| 0.2 mL | |
| Conical Bottom (V-well) | |
| Certified Quality |