missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperFi™ Buffers

Catalog No. 12356005
Encompass
Change view
Click to view available options
Color:
Colorless
Green
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
12356005 Green
12355005 Colorless
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12356005 Supplier Invitrogen™ Supplier No. 12356005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Invitrogen SuperFi Buffer is a 5X buffer solution optimized for use with Platinum SuperFi DNA Polymerase.

Invitrogen™ SuperFi™ Buffer is a 5X buffer solution optimized for use with Invitrogen™ Platinum™ SuperFi DNA Polymerase. The buffer contains 7.5 mM MgCl2 which provides 1.5 mM MgCl2 in the final reaction.

The SuperFi Green Buffer (Cat. No. 12356005) includes a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with the enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.

For applications that require PCR product analysis by absorbance or fluorescence excitation, we recommend using the colorless SuperFi™ Buffer. (12355005)

Specifications

Color Green
For Use With (Application) Hot-start PCR
Quantity 4 x 1.25 mL
Includes 5X SuperFi Green Buffer (4 x 1.25 mL)

Store at –5°C to –30°C.

For Research Use Only. Not for use in diagnostic procedures.