missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperBlock™ Blocking Buffer

SuperBlock Blocking Buffer is an optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis.

Supplier:  Thermo Scientific™ 37535

 View more versions of this product

Catalog No. PI37535


Only null left

Description

Description

SuperBlock Blocking Buffer is an optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis. Blocking micro-plates, membranes, or tissues with a SuperBlock buffer yields a high signal-to-noise ratio in most detection systems.

Features of SuperBlock Blocking Buffer include:

  • Blocking time—block membranes in 5 to 10 minutes and ELISA plates in 2 minutes
  • Applications—ELISA, western blotting, IHC; biotin-free for use with streptavidin system
  • Stability—store buffer at 4°C for one year; store blocked plates dry for up to 12 months

Blocking micro-plates, membranes, or tissues with SuperBlock buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin, or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilizing them for drying and storage for later use.

Specifications

Chemical Identifiers

ELISA
SuperBlock

Specifications

SuperBlock (TBS)
ELISA
SuperBlock
1X
Liquid
1 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Upon receipt store at 4°C. Product is shipped at ambient temperature.

SDS
Documents

Documents

Promotions

Promotions