Learn More
Metro Super Erecta EZ-ADD Shelf
Quickly add more storage to your Metro wire shelving with Super Erecta EZ-ADD Wire Shelves. The EZ-ADD Wire Shelf is designed to be easily added to an existing Metro wire shelving unit without dismantling the unit, minimizing the time and frustration associated with reconfiguring shelving.
Supplier:  Metro EZA1848NK3
 View more versions of this product
Catalog No. 11-120-540
Only null left
Explore available promotions
Description
Description
  • Compatible with Super Erecta, Super Adjustable Super Erecta, and Super Erecta Pro shelving.
  • Add/Remove an EZ-ADD shelf easily without dismantling a shelving unit.
  • Add more storage levels faster than conventional shelving.
  • Built-in Microban antimicrobial product protection fights the growth of bacteria, mold, and mildew that cause stains, odors, and product degradation.
  • 400 lbs. (181 kg) load capacity even distributed.
  • Can be used in many different applications, such as stationary and mobile shelving, security shelving, and track shelving solutions.
  • Metroseal finish has a 12-year limited warranty against rust and corrosion.
  • Unit load ratings are dependent on the number of Super Erecta shelves, Super Adjustable shelves, Super Erecta Pro shelves, and/or three-sided double snake frames used.
  • Please see Spec Sheet for additional details.
Specifications
Specifications
Product will be free from defects in material and workmanship under normal use, service and maintenance for a period of one (1) year from the date of shipment of the Product by Seller, unless otherwise specified by Seller in writing. Metroseal shelves and posts also carry a twelve (12) year warranty against rust formation and corrosion.
Add Shelf to Existing Shelving Unit
EZ-ADD Shelf and Mounting Hardware
5.4 kg
457.2 mm
Not Autoclavable
NSF Listed
Epoxy-coated Steel
Wire Shelf
48 x 18 in.
18 in.
11.3 lb.
Metro Super Erecta Shelving
Metroseal Green
Green
Product Suggestions
Product Suggestions