missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Super Concentrate Cleaner
Concentrated cleaning power for especially dirty floors. This purpose-built, eco-friendly cleaner tackles tough grime and adhesive residue for floor tape prep and cleaning.
$145.88 - $557.04
Specifications
| Product Type | Cleaner Concentrate |
|---|---|
| Color-coded | No |
| Form | Concentrated Liquid |
| Scent | Original |
| Sterility | Non-sterile |
Description
- Heavy-Duty Cleaning Power: Specifically formulated to cut through stubborn dirt, grime and old adhesive residue.
- Ensures Optimal Adhesion: Maximizes floor tape bond and durability by ensuring even the most challenging surfaces are clean.
- Green and Eco-Friendly: Safe, non-hazardous formula.
- Versatile Application: Effectively cleans a wide range of floor types.
- Just Add to Water: Single-use packet saves storage space and allows for quick mixing.
Specifications
| Cleaner Concentrate | |
| Concentrated Liquid | |
| Non-sterile |
| No | |
| Original |