missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Super Concentrate Cleaner
Description
- Heavy-Duty Cleaning Power: Specifically formulated to cut through stubborn dirt, grime and old adhesive residue.
- Ensures Optimal Adhesion: Maximizes floor tape bond and durability by ensuring even the most challenging surfaces are clean.
- Green and Eco-Friendly: Safe, non-hazardous formula.
- Versatile Application: Effectively cleans a wide range of floor types.
- Just Add to Water: Single-use packet saves storage space and allows for quick mixing.
Specifications
Specifications
| Product Type | Cleaner Concentrate |
| Color-coded | No |
| Form | Concentrated Liquid |
| Scent | Original |
| Sterility | Non-sterile |
| Diameter (English) | 9 in. |
| Height (English) | 6 in. |
| Quantity | 12 Ea. |
| Width (English) | 5 in. |