missing translation for 'onlineSavingsMsg'
Learn More

PIG™ Super Absorbent Sock

Catalog No. 18366622
Change view
Click to view available options
Dimensions (L x W):
42 x 3.25 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
18366622 42 x 3.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18366622 Supplier PIG™ Supplier No. PIG210

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Absorbent general-purpose sock to control high-volume machine leaks.

  • Formable socks hug corners and walls, surround higher-volume machine leaks and contain spills to keep workers safe
  • Cellulose filler is highly absorbent for big leaks and spills
  • Stitch-bonded, polypropylene skin resists bursting, reduces dust, and holds in liquid
  • Absorbs most common industrial liquids, oils, water, solvents, coolants, etc.
  • Not recommended for acids, bases or other corrosive liquids
  • 85% recycled cellulose filler
  • Wringable for reclaiming up to 65% of absorbed liquid
  • Can be incinerated after use to reduce waste or for fuels blending
  • 3.25 x 42 in.
  • Absorbs up to 1 gal. per sock
  • 24 socks per box

Specifications

Absorbancy 24 gal.
Material Cellulose, Polypropylene
Type Absorbent Sock
Disposable Single-use
Packaging Quantity 24/Cs.
Dimensions (L x W) 42 x 3.25 in.
Weight (English) 29.5 lb.
Weight (Metric) 13.4 kg