Learn More
MilliporeSigma™ Supelco™ Supeltex™ M-4 Capillary Ferrule, General Purpose, 0.10-0.25 mm Column I.D.
Supelco has refined the design of these graphite ferrules to provide a clean, sharp profile with minimal flash.
$126.91 - $425.21
Specifications
Diameter (Metric) Inner 0.4 mm
View More Specs
Products 2
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
111020905
View Documents Promotion Details
MilliporeSigma™ Supelco™
22498
10/Pk.
Each for $126.91
Only null left
111016348
View Documents Promotion Details
MilliporeSigma™ Supelco™
22480U
50/Pk.
Each for $425.21
Only null left
Description
Description
Characteristics: Seals at one-quarter turn past fingertight. Maximum sealing surface contact, reduced risk of column contamination at installation .Material Composition: flexible graphite. Maximum Temperature: 450°C
Specifications
Specifications
0.4 mm
Videos
SDS
Documents
Documents
Supeltex is a registered trademark of Sigma-Aldrich Co. LLC