Learn More
MilliporeSigma™ Supelco™ Supeltex™ M-4 Capillary Ferrule, General Purpose, 0.10-0.25 mm Column I.D.
Supelco has refined the design of these graphite ferrules to provide a clean, sharp profile with minimal flash.
Supplier:  MilliporeSigma™ Supelco™ 22480U
 View more versions of this product
Catalog No. 111016348
Only null left
Description
Description
Characteristics: Seals at one-quarter turn past fingertight. Maximum sealing surface contact, reduced risk of column contamination at installation .Material Composition: flexible graphite. Maximum Temperature: 450°C
Specifications
Chemical Identifiers
0.4 mm
Specifications
0.4 mm
50/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Supeltex is a registered trademark of Sigma-Aldrich Co. LLC