missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Supelpure™-O Oxygen/Moisture Trap

Catalog No. 111016414
Change view
Click to view available options
Capacity (Metric):
120 cc
750 cc
Size:
1/2 in.
1/4 in.
1/8 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Size
111016414 120 cc 1/8 in.
111016413 120 cc 1/4 in.
111003761 750 cc 1/4 in.
111003770 750 cc 1/2 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 111016414 Supplier MilliporeSigma™ Supelco™ Supplier No. 22449

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Oxygen-removing catalysts that can reduce oxygen to less than 2 ppb when the level in the incoming gas does not exceed 10 ppm

Specifications

Capacity (Metric) 120 cc
Product Line Supelpure
Type Oxygen/Moisture Trap
For Use With (Application) Removing Oxygen and Water Vapor From Incoming Gas
Size 1/8 in.
Length (English) 11.125 in.
Length (Metric) 28.2 cm
Diameter (English) Outer 1 3/8 in.
Diameter (Metric) Outer 35 mm

Supelpure is a registered trademark of Sigma-Aldrich Co. LLC