missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Supelcarb™ HC Hydrocarbon Trap

Catalog No. 111004070
Change view
Click to view available options
Capacity (Metric):
120 cc
750 cc
Size:
1/2 in.
1/4 in.
1/8 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Size
111004070 120 cc 1/4 in.
111004069 120 cc 1/8 in.
111003480 750 cc 1/2 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111004070 Supplier MilliporeSigma™ Supelco™ Supplier No. SGMA24449

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The best trap available for removing hydrocarbons and other organics from carrier, fuel, air, and other gas streams

  • Primarily used for trapping hydrocarbons
  • Used in GC for carrier gas purification
  • Removes organics from carrier gases, air, and hydrogen
  • Adsorbent has twice the trapping ability of activated charcoal

Specifications

Capacity (Metric) 120 cc
Product Line Supelcarb
Type Hydrocarbon Trap
For Use With (Application) Removing Hydrocarbons and Other Organics From Carrier, Fuel, Air, and Other Gas Streams
Size 1/4 in.
Length (English) 11.125 in.
Length (Metric) 28.2 cm
Diameter (English) Outer 1 3/8 in.
Diameter (Metric) Outer 35 mm

Supelcarb is a registered trademark of Sigma-Aldrich Co. LLC