missing translation for 'onlineSavingsMsg'
Learn More

Supel™-Inert Multi-Layer Foil Gas Sampling Bags, Sold by MilliporeSigma™ Supelco™

Catalog No. 111010085
Change view
Click to view available options
Fill Volume:
0.6 L
1 L
2 L
5 L
10 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fill Volume
111010085 5 L
111010610 0.6 L
111020864 1 L
111010437 2 L
111009900 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 111010085 Supplier Merck Emd Millipore Supplier No. 30228U
Only null left

Ideal for sampling low molecular weight compounds. Two outer layers of aluminum film provide a great barrier to gases permeating through the walls of the bag.

  • Proven low-bleed Thermogreen™ LB-2 Septa installed in the valve fitting
  • Chemically inert with moisture and light protection
  • Not recommended for low ppm VOCs due to background levels

Specifications

Description Chemically Inert with Light/Moisture Protection
Certifications/Compliance NIOSH 6603, OSHA ID-172 (ID-210)
Closure Type Screw Cap Valve (SCV)
For Use With (Application) Sampling Low Molecular Weight Compounds Such as Carbon Monoxide (CO), Carbon Dioxide (CO2) and Permanent Gases. Also Suitable for Sampling Sulfur Compounds Such as Hydrogen Sulfide (H2S), Carbon Disulfide (CS2) and Mercaptans.
Fill Volume 5 L
Film Description Multi-layer foil composed of two outer layers of aluminum film
No. per Pack 10
Port Detail Thermogreen™ LB-2 Septa
Product Line Supel
Resistant To Chemicals
Length (English) Document Pouch 14 in.
Width (English) Document Pouch 12 in.
Product Type Multi-layer Foil Gas Sampling Bags
Show More Show Less

Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC