missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Supel™-Inert Multi-Layer Foil Gas Sampling Bags, Sold by MilliporeSigma™ Supelco™
Ideal for sampling low molecular weight compounds. Two outer layers of aluminum film provide a great barrier to gases permeating through the walls of the bag.
Supplier: Merck Emd Millipore 30229U
Description
- Proven low-bleed Thermogreen™ LB-2 Septa installed in the valve fitting
- Chemically inert with moisture and light protection
- Not recommended for low ppm VOCs due to background levels
Chemical Identifiers
| Chemically Inert with Light/Moisture Protection | |
| Screw Cap Valve (SCV) | |
| Multi-layer foil composed of two outer layers of aluminum film | |
| Thermogreen™ LB-2 Septa | |
| Chemicals |
| NIOSH 6603, OSHA ID-172 (ID-210) | |
| Sampling Low Molecular Weight Compounds Such as Carbon Monoxide (CO), Carbon Dioxide (CO2) and Permanent Gases. Also Suitable for Sampling Sulfur Compounds Such as Hydrogen Sulfide (H2S), Carbon Disulfide (CS2) and Mercaptans. | |
| 10 | |
| Supel |
Specifications
| Chemically Inert with Light/Moisture Protection | |
| Screw Cap Valve (SCV) | |
| 10 L | |
| 10 | |
| Supel | |
| 21 in. | |
| Multi-layer Foil Gas Sampling Bags |
| NIOSH 6603, OSHA ID-172 (ID-210) | |
| Sampling Low Molecular Weight Compounds Such as Carbon Monoxide (CO), Carbon Dioxide (CO2) and Permanent Gases. Also Suitable for Sampling Sulfur Compounds Such as Hydrogen Sulfide (H2S), Carbon Disulfide (CS2) and Mercaptans. | |
| Multi-layer foil composed of two outer layers of aluminum film | |
| Thermogreen™ LB-2 Septa | |
| Chemicals | |
| 12 in. |
Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC