missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Supel™-Inert Multi-Layer Foil Gas Sampling Bag

Catalog No. 111010622
Change view
Click to view available options
Capacity (Metric):
0.6 L
1 L
2 L
10 L
Dimensions (L x W):
6 x 6 in.
9 x 7 in.
11 x 9 in.
21 x 12 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Dimensions (L x W)
111010622 0.6 L 6 x 6 in.
111010596 1 L 9 x 7 in.
111010438 2 L 11 x 9 in.
111009920 10 L 21 x 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 111010622 Supplier MilliporeSigma™ Supelco™ Supplier No. 30281U

missing translation for 'validMainframeMsg'

Ideal for sampling low molecular weight compounds such as carbon monoxide (CO), carbon dioxide (CO2) and permanent gases

Supel™-Inert multi-layer foil gas sampling bags with Thermogreen™ LB-2 septa are ideal for sampling low molecular weight compounds such as carbon monoxide (CO), carbon dioxide (CO2) and permanent gases. They are also suitable for sampling sulfur compounds such as hydrogen sulfide (H2S), carbon disulfide (CS2) and mercaptans. Key Features and Benefits: Multi-layer foil composed of two outer layers of aluminum film providing a great barrier to gases permeating through walls of the bag. Proven low-bleed Thermogreen™ LB-2 septa installed in the valve fitting. Chemically inert with moisture and light protection. Not recommended for low ppm VOCs due to background levels (we recommend the Supel™Inert PVDF Tedlar alternative film for VOCs). Two valve choices available: Screw Cap Valve (SCV) and Push/Pull Lock Valve (PLV). 4 sizes: 1 L, 2 L, 5 L, and 10 L

Specifications

Includes Push/Pull Lock Valve (PLV)
Material Multi-layer Foil
Product Line Supel
Quantity 10/Pk.
Dimensions (L x W) 6 x 6 in.
Capacity (Metric) 0.6 L

Supel and Thermogreen are registered trademarks of Sigma-Aldrich Co. LLC