missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SUOX Monoclonal Antibody (K92029_4B1), Invitrogen™
Mouse Monoclonal Antibody
$1,740.58 - $2,926.56
Specifications
| Antigen | SUOX |
|---|---|
| Clone | K92029_4B1 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: PVPNLDPDTY RLHVVGAPGG QSLSLSLDDL HNFPRYEITV TLQCAGNRRS EMTQVKEVKG LEWRTGAIST ARWAGARLCD VLAQAGHQLC ETEAHVCFEG LDSDPTGTAY GASIPLARAM DPEAEVLLAY EMNGQPLPRD HGFPVRVVVP GVVGARHVKW LGRVSVQPEE SYSHWQRRDY KGFSPSVDWE TVDFDSAPSI QELPVQSAIT EPRDGETVES GEVTIKGYAW SGGGRAVIRV DVSLDGGLTW QVAKLDGEEQ RPRKAWAWRL WQLKAPVPAG QKELNIVCKA VDDGYNVQPD TVAPIWNLRG VLSNAWHRVH
SUOX Monoclonal antibody specifically detects SUOX in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
| SUOX | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| SO; sulfite oxidase; sulfite oxidase, mitochondrial; Suox | |
| Recombinant human SUOX (immunogen range is 221-540). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| SUOX |
| K92029_4B1 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P51687 | |
| 6821 | |
| IgG1 | |
| Protein A | |
| Antibody |