missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SUMO-3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 401900
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
401900 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 401900 Supplier Invitrogen™ Supplier No. 401900
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

SUMO-3 Polyclonal Antibody for Western Blot

Small ubiquitin-related modifier proteins are members of the ubiquitin-like protein family. SUMO-3 regulates Amyloid beta generation and may be critical in the onset or progression of Alzheimer's disease. SUMO-2/3 forms poly-. SUMO-1, 2, and 3 proteins localize to the nuclear membrane, nuclear bodies, and cytoplasm, respectively. SUMOylation is essential for mammalian cell cycle regulation, SUMO-2/3 localizes to centromeres and condensed chromosomes where binds CENP-E.

Specifications

Antigen SUMO-3
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene SUMO3
Gene Accession No. P55854
Gene Alias 2810014B19Rik; D10Ertd345e; small ubiquitin like modifier 3; small ubiquitin-like modifier 3; small ubiquitin-related modifier 3; SMT3 (supressor of mif two, 3) homolog 1; SMT3 homolog 1; SMT3 suppressor of mif two 3 homolog 1; SMT3 suppressor of mif two 3 homolog 3; SMT3 suppressor of mif two 3 homolog 3 (yeast); Smt3A; Smt3B; SMT3H1; SUMO-2; Sumo3; SUMO-3; Ubiquitin-like protein SMT3A; ubiquitin-like protein SMT3B
Gene Symbols SUMO3
Host Species Rabbit
Immunogen Synthetic peptide derived from the C-terminal region of the human SUMO-3 protein.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6612
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less