missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SUMF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. pipa554961
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554961 Supplier Invitrogen™ Supplier No. PA554961
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: ATSMVQLQGG RFLMGTNSPD SRDGEGPVRE ATVKPFAIDI FPVTNKDFRD FVREKKYRTE AEMFGWSFVF EDFVSDELRN KATQPMKSVL WWLP Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 77%.

Specifications

Antigen SUMF2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SUMF2
Gene Accession No. Q8NBJ7
Gene Alias 2610040F05Rik; C-alpha-formyglycine-generating enzyme 2; C-alpha-formylglycine-generating enzyme 2; DKFZp566I1024; DKFZp686I1024; DKFZp686L17160; DKFZp781L1035; FGE2; Inactive C-alpha-formylglycine-generating enzyme 2; MGC99485; Paralog of formylglycine-generating enzyme; paralog of the formylglycine-generating enzyme; pFGE; PSEC0171; sulfatase modifying factor 2; sulfatase-modifying factor 2; Sumf2; UNQ1968/PRO4500
Gene Symbols SUMF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SUMF2. Recombinant protein control fragment (Product #RP-92716).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25870
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less