missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SULT2B1 Monoclonal Antibody (K06341_3C4), Invitrogen™

Catalog No. MA556328
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556328 200 μL
MA555948 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556328 Supplier Invitrogen™ Supplier No. MA556328
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: EAFDRAYRKQ MRGMPTFPWD EDPEEDGSPD PEPSPEPEPK PSLEPNTSLE REPRPNSSPS PSPGQASETP

Cytosolic sulfotransferases (STs or SULTs) catalyze the sulfate conjugation of many drugs, xenobiotic compounds, hormones, and neurotransmitters. Her et al. (1998) identified an EST with homology to ST enzymes. By PCR with human placental and prostate cDNA as template, they isolated 2 alternatively spliced cDNAs, identical throughout most of their sequences, but having different 5-prime ends. The shorter cDNA, SULT2B1a, encodes a protein of 350 amino acids; the longer cDNA, SULT2B1b, encodes a 365-amino acid protein. Genomic PCR analysis revealed that the gene encoding both cDNAs, SULT2B1, contains 7 exons, with 2 alternative first exons being used to generate SULT2B1a and SULT2B1b. Northern blot analysis revealed that the SULT2B1 gene is expressed as a 1.4-kb transcript predominantly in prostate, placenta, and trachea.

Specifications

Antigen SULT2B1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K06341_3C4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene SULT2B1
Gene Accession No. O00204
Gene Alias AI326997; alcohol sulfotransferase; BB173635; HSST2; Hydroxysteroid sulfotransferase 2; RGD:1308882}; ST2B1; sulfotransferase 2B; sulfotransferase 2B1; Sulfotransferase family 2B member 1; sulfotransferase family cytosolic 2B member 1; sulfotransferase family, cytosolic, 2B, member 1; SULT2B; Sult2b1; sult2b1 {ECO:0000312; SULT2B1a
Gene Symbols SULT2B1
Host Species Mouse
Immunogen Recombinant human SULT2B1 (immunogen range is 291-360).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6820
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less