missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SULT1A3/SULT1A4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116182
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116182 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116182 Supplier Invitrogen™ Supplier No. PA5116182
Only null left

Rabbit Polyclonal Antibody

SULT1A3/SULT1A4 Polyclonal Antibody for Western Blot, IHC (P)

Found in a segmental duplication on p arm of chromosome 16 giving rise to two identical copies of this gene sharing exons with SLX1A and SLX1B. The ORFs of SULT1A3 and SULT1A4 differ with only a single nucleotide change that does not alter the encoded amino acid. It is not possible to determine whether any individual polymorphism is present within SULT1A3 or SULT1A4 (PubMed:15358107).

Specifications

Antigen SULT1A3/SULT1A4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SULT1A3
Gene Accession No. P0DMM9, P0DMN0
Gene Alias aryl sulfotransferase 1A3/1A4; Catecholamine-sulfating phenol sulfotransferase; dopamine-specific sulfotransferase; HAST; HAST3; Monoamine-sulfating phenol sulfotransferase; monoamine-sulfating phenosulfotransferase; M-PST; phenol sulfotransferase 1A5; placental estrogen sulfotransferase; SLX1B-SULT1A4; ST1A3; ST1A3/ST1A4; ST1A4; ST1A5; STM; sulfokinase; sulfotransferase 1A3; Sulfotransferase 1A3/1A4; Sulfotransferase 1A4; sulfotransferase family 1A member 3; sulfotransferase family 1A member 4; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 3; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 4; sulfotransferase, monoamine-preferring; SULT1A3; SULT1A4; thermolabile (monoamine, M form) phenol sulfotransferase; thermolabile phenol sulfotransferase; TL-PST
Gene Symbols SULT1A3, SULT1A4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 71-99 amino acids from the N-terminal region of human SULT1A3/SULT1A4.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 445329, 6818
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less