missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (±)-Sulpiride

Catalog No. AC463481000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463481000 100 g
AC463480050 5 g
AC463480250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC463481000 Supplier Thermo Scientific Chemicals Supplier No. 463481000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 15676-16-1 | C15H23N3O4S | 341.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15676-16-1
Molecular Formula C15H23N3O4S
Molecular Weight (g/mol) 341.43
MDL Number MFCD00055061
InChI Key BGRJTUBHPOOWDU-UHFFFAOYNA-N
IUPAC Name N-[(1-ethylpyrrolidin-2-yl)methyl]-2-methoxy-5-sulfamoylbenzamide
SMILES CCN1CCCC1CNC(=O)C1=CC(=CC=C1OC)S(N)(=O)=O

Specifications

Melting Point 185.0°C
Quantity 100 g
Formula Weight 341.43
Chemical Name or Material (±)-Sulpiride
EINECSNumber 239-753-7

RUO – Research Use Only