missing translation for 'onlineSavingsMsg'
Learn More

Sulindac 98.0+%, TCI America™

Catalog No. p-7137873
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U009910G 10 g
U009950G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U009910G Supplier TCI America Supplier No. U009910G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 38194-50-2
Molecular Formula C20H17FO3S
Molecular Weight (g/mol) 356.411
MDL Number MFCD00599589
InChI Key MLKXDPUZXIRXEP-MFOYZWKCSA-N
Synonym sulindac, clinoril, arthrocine, sulindac sulfoxide, sulindaco, sulindacum, mobilin, arthrobid, klinoril, sulindacum inn-latin
PubChem CID 1548887
ChEBI CHEBI:9352
IUPAC Name 2-[(3Z)-6-fluoro-2-methyl-3-[(4-methylsulfinylphenyl)methylidene]inden-1-yl]acetic acid
SMILES CC1=C(C2=C(C1=CC3=CC=C(C=C3)S(=O)C)C=CC(=C2)F)CC(=O)O

Specifications

Melting Point 184°C
Quantity 10 g
UN Number 2811
Formula Weight 356.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sulindac
RTECSNumber NK8226000
TSCA No