Learn More
Thermo Scientific Chemicals Sulindac
CAS: 38194-50-2 | C20H17FO3S, C20H17FO3S | 356.41 g/mol
$138.16 - $524.37
Chemical Identifiers
| CAS | 38194-50-2 |
|---|---|
| Molecular Formula | C20H17FO3S |
| Synonym | cis-5-Fluoro-2-methyl-1-[4-(methylsulfinyl)benzylidene]indene-3-acetic acid; MK-231 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ6177206
|
Thermo Scientific Chemicals
J6177206 |
5 g |
Each for $138.16
|
|
|||||
|
AAJ6177214
|
Thermo Scientific Chemicals
J6177214 |
25 g |
Each for $524.37
|
|
|||||
Description
Sulindac inhibits the NF-κB pathway. Sulindac is a non-steroidal anti-inflammatory drug. Sulindac reduces the level and activity of several cyclin-dependent kinases and induces apoptosis. It is an antitumor agent. Sulindac is an inhibitor of Cox-1 and Cox-2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase-1 and induces apoptosis in HT-29 cells.Chemical Identifiers
| 38194-50-2 | |
| C20H17FO3S | |
| cis-5-Fluoro-2-methyl-1-[4-(methylsulfinyl)benzylidene]indene-3-acetic acid; MK-231 |
Specifications
| Yellow | |
| A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase-1 and induces apoptosis in HT-29 cells | |
| C20H17FO3S | |
| cis-5-Fluoro-2-methyl-1-[4-(methylsulfinyl)benzylidene]indene-3-acetic acid; MK-231 | |
| Signal Transduction Reagent |
| -30°C to -10°C | |
| 38194-50-2 | |
| 356.41g/mol | |
| 182°C to 185°C | |
| Powder |
Safety and Handling
EINECSNumber : 253-819-2
Recommended Storage : Store at -20°C
RUO – Research Use Only