missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sulindac

Catalog No. AAJ6177214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6177214 25 g
AAJ6177206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6177214 Supplier Thermo Scientific Chemicals Supplier No. J6177214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 38194-50-2 | C20H17FO3S, C20H17FO3S | 356.41 g/mol

Sulindac inhibits the NF-κB pathway. Sulindac is a non-steroidal anti-inflammatory drug. Sulindac reduces the level and activity of several cyclin-dependent kinases and induces apoptosis. It is an antitumor agent. Sulindac is an inhibitor of Cox-1 and Cox-2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase-1 and induces apoptosis in HT-29 cells.

Chemical Identifiers

CAS 38194-50-2
Molecular Formula C20H17FO3S
Synonym cis-5-Fluoro-2-methyl-1-[4-(methylsulfinyl)benzylidene]indene-3-acetic acid; MK-231

Specifications

Quantity 25 g
Description Yellow
Content And Storage -30°C to -10°C
For Use With (Application) A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase-1 and induces apoptosis in HT-29 cells
Formula Weight 356.41g/mol
Melting Point 182°C to 185°C
Product Type Signal Transduction Reagent
Form Powder

RUO – Research Use Only