missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sulindac

CAS: 38194-50-2 | C20H17FO3S, C20H17FO3S | 356.41 g/mol

Supplier:  Thermo Scientific Chemicals J6177206

 View more versions of this product

Catalog No. AAJ6177206


Only null left

Description

Description

Sulindac inhibits the NF-κB pathway. Sulindac is a non-steroidal anti-inflammatory drug. Sulindac reduces the level and activity of several cyclin-dependent kinases and induces apoptosis. It is an antitumor agent. Sulindac is an inhibitor of Cox-1 and Cox-2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase-1 and induces apoptosis in HT-29 cells.
Specifications

Specifications

5 g
-30°C to -10°C
38194-50-2
356.41g/mol
182°C to 185°C
Powder
Yellow
A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase-1 and induces apoptosis in HT-29 cells
C20H17FO3S
cis-5-Fluoro-2-methyl-1-[4-(methylsulfinyl)benzylidene]indene-3-acetic acid; MK-231
Signal Transduction Reagent

Chemical Identifiers

38194-50-2
C20H17FO3S
cis-5-Fluoro-2-methyl-1-[4-(methylsulfinyl)benzylidene]indene-3-acetic acid; MK-231
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 253-819-2

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only