missing translation for 'onlineSavingsMsg'
Learn More

Sulfurous Acid, 6% ACS Reagent, Reagents

Catalog No. 01338160
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338160 1 L
01338159 2.5 L
01338158 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01338160 Supplier Labchem Inc Supplier No. C11155001C
Only null left

Sulfurous Acid, 6% ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7782-99-2 , 7732-18-5
Molecular Formula H2O3S
Molecular Weight (g/mol) 82.07
InChI Key LSNNMFCWUKXFEE-UHFFFAOYSA-N
Synonym Sulfur dioxide solution, Sulphurous Acid
IUPAC Name sulfurous acid
SMILES OS(O)=O

Specifications

pH <7
Color Colorless
Boiling Point 35°C
Packaging Glass Bottle
Specific Gravity 1.03
Quantity 1 L
Physical Form Liquid
Grade ACS
Chemical Name or Material Sulfurous Acid, 6% ACS Reagent