missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, TraceSELECT™, for trace analysis,≥95%, Honeywell Fluka™

Catalog No. p-7121630
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60047676 500 mL Plastic Bottle
60047674 1 L Plastic Bottle
60047675 2.5 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60047676 Supplier Honeywell Inc Supplier No. 84716500ML
Only null left

TraceSELECT™, for trace analysis,≥95%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Boiling Point 310°C
CAS Min % ≥50.0000%, ≥1.0000%
CAS Max % ≤100.0000%, <5.0000%
Density 1.840 g/mL at 25 °C(lit.)
Packaging Plastic Bottle
Vapor Pressure 1 mmHg (146°C)
Quantity 500 mL
Linear Formula H2SO4
UN Number UN1830
Beilstein 2037554
Formula Weight 98.08
Percent Purity ≥95%
Chemical Name or Material Sulfuric acid, TraceSELECT™, for trace analysis,≥95%
Show More Show Less
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
ShelfLife 900 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Sulfuric acid
EINECSNumber 231-639-5
RTECSNumber 231-639-5
Recommended Storage Room Temp