missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 10% (w/w), Ricca Chemical
$122.38 - $122.38
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O4S |
| Molecular Weight (g/mol) | 98.07 |
| MDL Number | MFCD00064589 |
| InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
| Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
| PubChem CID | 1118 |
| ChEBI | CHEBI:26836 |
| IUPAC Name | sulfuric acid |
| SMILES | OS(O)(=O)=O |
Chemical Identifiers
| 7732-18-5 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| 1118 | |
| sulfuric acid |
| H2O4S | |
| MFCD00064589 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| CHEBI:26836 | |
| OS(O)(=O)=O |
Specifications
| Colorless | |
| 89.0,9.0 | |
| 91.0,11.0 | |
| Aqueous Solution | |
| H2O4S | |
| MFCD00064589 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 | |
| 10% W/W | |
| Sulfuric Acid Solution |
| 7732-18-5,7664-93-9 | |
| 10% (w/w) | |
| Natural Poly Bottle | |
| 1 L | |
| Liquid | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| OS(O)(=O)=O | |
| 98.07 | |
| CHEBI:26836 | |
| Laboratory |