missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid Solution (4N), Fisher Chemical™

Catalog No. SA818500
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Poly Bottle
PolyPac™
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SA818500 500 mL Poly Bottle
SA8181 1 L Poly Bottle
SA8184 4 L Poly Bottle
SA81820 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SA818500 Supplier Thermo Fisher Scientific Supplier No. SA818500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Standardized to pH 4.4 at 25°C against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 0.3
Melting Point 10°C
Boiling Point 290°C
Concentration 3.95 to 4.05
Density 1.5g/cm³
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure <0.001 mmHg at 20°C
Decomposition Information 340°C
Quantity 500 mL
Physical Form Liquid
UN Number 2796
Chemical Name or Material Sulfuric Acid Solution, 4N
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Causes severe burns by all exposure routes. Water reactive. Contact with combustible material may cause fire. Use personal protective equipment. Use only under a chemical fume hood. Do not allow contact with water. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:3
Flammability:0
Instability:2
DOTInformation DOT Class 8, : Corrosive