missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid Solution (10N/Certified), Fisher Chemical

Catalog No. sa2001
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SA2001 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SA2001 Supplier Thermo Fisher Scientific Formerly Fairlawn Supplier No. FLSA2001
Only null left

Standardized to pH 4.4 at 25°C against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 0.03
Melting Point 11°C
Boiling Point 105°C
Concentration 9.95 to 10.05
Identification Pass Test
Packaging Glass Bottle
Traceability to NIST LOT 84L Potassium acid phthalate
Vapor Pressure <0.00120 mmHg
Quantity 1 L
Physical Form Liquid
UN Number 2796
Grade Certified
Concentration or Composition (by Analyte or Components) 10N
Chemical Name or Material Sulfuric Acid Solution, 10N
Show More Show Less