missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid Solution, 0.5N (N/2) (Certified), Fisher Chemical™

Catalog No. p-20576
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Poly Bottle
Catalog No. Quantity Packaging
SA2151 1 L Poly Bottle
SA2154 4 L Poly Bottle
2 options

Catalog No. SA2151

Supplier: Thermo Fisher Scientific SA215 1

Only null left

Standardized to pH 4.4 at 25°C against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 1.0
Concentration 0.498 to 0.500N
Identification Pass Test
Packaging Poly Bottle
Decomposition Information 340°C
Quantity 1 L
Physical Form Liquid
UN Number 2796
Grade Certified
Concentration or Composition (by Analyte or Components) 0.5N (N/2)
Chemical Name or Material Sulfuric Acid Solution, 0.5N (N/2)
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Causes severe eye and skin irritation with possible burns. May be harmful if inhaled. Corrosive to metals. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:3
Flammability:0
Instability:1
DOTInformation DOT Class 8, : Corrosive