missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid solution, Volumetric, Reag. Ph. Eur., 0.5Â M H2SO4 (1.0N), Honeywell Fluka™
Volumetric, Reag. Ph. Eur., 0.5Â M H2SO4 (1.0N)
$198.23 - $482.75
Chemical Identifiers
| CAS | 7664-93-9 |
|---|---|
| Molecular Formula | H2O4S |
| Molecular Weight (g/mol) | 98.07 |
| MDL Number | MFCD00064589 |
| InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
| PubChem CID | 1118 |
| ChEBI | CHEBI:26836 |
| IUPAC Name | sulfuric acid |
| SMILES | OS(O)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
6001613
|
Honeywell Inc
3535425L |
2.5 L | Plastic Bottle |
Each for $198.23
|
|
||||
|
60046574
|
Honeywell-Fluka
353546X1L |
6 x 1 L | Plastic Bottle |
Case of 6 Each for $473.78
|
|
||||
|
6001611
|
Honeywell-Fluka
3535410LVP |
10 L | VOLPAC™ CONTAINER |
Each for $482.75
|
|
||||
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
| 7664-93-9 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| CHEBI:26836 | |
| OS(O)(=O)=O |
| H2O4S | |
| MFCD00064589 | |
| 1118 | |
| sulfuric acid |
Specifications
| 7664-93-9 | |
| Plastic Bottle | |
| H2O4S | |
| MFCD00064589 | |
| 2037554 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 | |
| 98.08g/mol | |
| Sulfuric acid solution |
| 0.5M | |
| 2.5 L | |
| H2SO4 | |
| UN3264 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| OS(O)(=O)=O | |
| 98.07 | |
| CHEBI:26836 | |
| Reagent Ph. Eur. |
Safety and Handling
ShelfLife : 1080 days from date of manufacture
DOTInformation : Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Corrosive liquid, acidic, inorganic, n.o.s.
Recommended Storage : Room Temp