missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid solution, Volumetric, Reag. Ph. Eur., 0.5Â M H2SO4 (1.0N), Honeywell Fluka™
Volumetric, Reag. Ph. Eur., 0.5Â M H2SO4 (1.0N)
Supplier: Honeywell-Fluka 353546X1L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
| Sulfuric acid solution | |
| 7664-93-9,7732-18-5 | |
| ≥90.0000%,≥1.0000% | |
| 1.030 g/cm3 | |
| Plastic Bottle | |
| H2O4S | |
| MFCD00064589 | |
| 2037554 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 | |
| 98.08 |
| Volumetric, Reag. Ph. Eur., 0.5Â M H2SO4 (1.0N) | |
| 100°C | |
| 0.5Â M H2SO4 (1.0N) | |
| ≤100.0000%,<5.0000% | |
| 6 x 1 L | |
| H2SO4 | |
| UN3264 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| OS(O)(=O)=O | |
| 98.07 | |
| CHEBI:26836 | |
| Reagent Ph. Eur. |
Chemical Identifiers
| 7664-93-9 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| CHEBI:26836 | |
| OS(O)(=O)=O |
| H2O4S | |
| MFCD00064589 | |
| 1118 | |
| sulfuric acid |
Safety and Handling
ShelfLife : 1080 days from date of manufacture
DOTInformation : Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Corrosive liquid, acidic, inorganic, n.o.s.
Recommended Storage : Room Temp